Alfa Aesar™ Tetra-n-butylammonium acetate, 1.0M aq. soln.

Manufacturer: Alfa Aesar™ B2477336

Quantity 500g

 View more versions of this product

Catalog No. AAB2477336

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 10534-59-5
    Molecular Formula C18H39NO2
    Linear Formula [CH3(CH2)3]4N(O2CCH3)
    Formula Weight 301.51
    MDL Number MFCD00043208
    Beilstein 3599376
    Chemical Name or Material Tetra-n-butylammonium acetate

    Specifications

    Formula Weight 301.51
    TSCA TSCA
    Linear Formula [CH3(CH2)3]4N(O2CCH3)
    Beilstein 3599376
    Health Hazard 1 Warning
    Health Hazard 3 GHS P Statement
    P261-P280-P305+P351+P338-P304+P340-P405-P501a
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    Wear protective gloves/protective clothing/eye protection/face protection.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
    IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.
    Boiling Point 100°C
    Concentration 1.0M aq. soln.
    Quantity 500g
    Molecular Formula C18H39NO2
    MDL Number MFCD00043208
    CAS 10534-59-5
    Health Hazard 2 GHS H Statement
    H315-H319-H335
    Causes skin irritation.
    Causes serious eye irritation.
    May cause respiratory irritation.
    Chemical Name or Material Tetra-n-butylammonium acetate

    GHS H Statement
    H315-H319-H335
    Causes skin irritation.
    Causes serious eye irritation.
    May cause respiratory irritation.

    GHS P Statement
    P261-P280-P305+P351+P338-P304+P340-P405-P501a
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    Wear protective gloves/protective clothing/eye protection/face protection.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
    IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.

    Warning

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit