Alfa Aesar™ Tetra-n-butylammonium bromide, 98+%

Manufacturer: Alfa Aesar™ A1024936

Quantity 500g

 View more versions of this product

Catalog No. AAA1024936

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 1643-19-2
    Molecular Formula C16H36BrN
    Linear Formula [CH3(CH2)3]4NBr
    Formula Weight 322.38
    MDL Number MFCD00011633
    Beilstein 3570983
    Synonym TBAB; Aliquat 100
    Chemical Name or Material Tetra-n-butylammonium bromide

    Specifications

    Formula Weight 322.38
    Health Hazard 3 GHS P Statement
    P261-P280-P305+P351+P338-P304+P340-P405-P501a
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    Wear protective gloves/protective clothing/eye protection/face protection.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
    Continue rinsing.
    IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.
    Molecular Formula C16H36BrN
    CAS 1643-19-2
    Synonym TBAB; Aliquat 100
    Quantity 500g
    MDL Number MFCD00011633
    Health Hazard 1 Warning
    Health Hazard 2 GHS H Statement
    H315-H319-H335
    Causes skin irritation.
    Causes serious eye irritation.
    May cause respiratory irritation.
    Melting Point 101° to 105°C
    Odor Amine-like
    TSCA TSCA
    Linear Formula [CH3(CH2)3]4NBr
    Beilstein 3570983
    Chemical Name or Material Tetra-n-butylammonium bromide
    Assay ≥98%

    Description

    Applications
    Phase transfer catalyst Tetra-n-butylammonium bromide is used as a phase transfer catalyst for the preparation of lactones by cyclization of potassium salt of µ-bromocarboxylic acids and beta-lactones from beta-bromoacid by cyclization. It is also involved in the Curtius rearrangement, N-alkylation of aromatic carboxamides. It plays an important role as an ionic liquid in the regioselective O-alkylation reactions. It is also used in the preparation of polymer solar cells and single component green-light emitting electrochemical cells.

    Solubility
    Soluble in water and ethanol.

    Notes
    Hygroscopic. Keep the container tightly closed in a dry and well-ventilated place. Incompatible with strong oxidizing agents.

    GHS H Statement
    H315-H319-H335
    Causes skin irritation.
    Causes serious eye irritation.
    May cause respiratory irritation.

    GHS P Statement
    P261-P280-P305+P351+P338-P304+P340-P405-P501a
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    Wear protective gloves/protective clothing/eye protection/face protection.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
    Continue rinsing.
    IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.

    Warning

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit