Alfa Aesar™ Tetra-n-butylammonium dichromate, 99%

Manufacturer: Alfa Aesar™ H5520506

Quantity 5g

 View more versions of this product

Catalog No. AAH5520506

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 56660-19-6
    Molecular Formula C32H72Cr2N2O7
    Linear Formula ([CH3(CH2)3]4N)2(Cr2O7)
    Formula Weight 700.92
    MDL Number MFCD00013104
    UN Number UN1479
    Synonym Bis(tetra-n-butylammonium) dichromate; TBADC
    Chemical Name or Material Tetra-n-butylammonium dichromate

    Specifications

    Formula Weight 700.92
    MDL Number MFCD00013104
    CAS 56660-19-6
    UN Number UN1479
    Synonym Bis(tetra-n-butylammonium) dichromate; TBADC
    Health Hazard 1 Danger
    Health Hazard 3 GHS P Statement
    P221-P210-P220-P363-P405-P501a
    Take any precaution to avoid mixing with combustibles.
    Keep away from heat/sparks/open flames/hot surfaces. - No Smoking.
    Keep/Store away from clothing/combustible materials.
    Wash contaminated clothing before reuse.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.
    Quantity 5g
    Molecular Formula C32H72Cr2N2O7
    Linear Formula ([CH3(CH2)3]4N)2(Cr2O7)
    Health Hazard 2 GHS H Statement
    H272-H350-H317
    May intensify fire; oxidizer.
    May cause cancer.
    May cause an allergic skin reaction.
    DOT Information Transport Hazard Class: 5.1; Packing Group: III; Proper Shipping Name: OXIDIZING SOLID, N.O.S.
    Melting Point 139° to 143°C
    Chemical Name or Material Tetra-n-butylammonium dichromate
    Assay 99%

    Description

    Applications
    Reagent for preparation of nitrophenol derivatives and quinones under aprotic reaction conditions, conversion of oximes to carbonyl compounds under microwave irradiation, catalyst precursor for catalytic gas phase dehydration of acetic acid to ketene, crosslinking agent.

    Solubility
    Soluble in water.

    Notes
    Keep container tightly closed. Store away from oxidizing agents.

    GHS H Statement
    H272-H350-H317
    May intensify fire; oxidizer.
    May cause cancer.
    May cause an allergic skin reaction.

    GHS P Statement
    P221-P210-P220-P363-P405-P501a
    Take any precaution to avoid mixing with combustibles.
    Keep away from heat/sparks/open flames/hot surfaces. - No Smoking.
    Keep/Store away from clothing/combustible materials.
    Wash contaminated clothing before reuse.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.

    Danger

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit