Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ Tetra-n-butylammonium hydroxide, 55% w/w aq. soln.

As a reagent for ion-pair extractions

Manufacturer:  Alfa Aesar™ 16198A1

Quantity 1kg

 View more versions of this product

Catalog No. AA16198A1


  Check Availability
Add to cart

Specifications Description & Specifications

Chemical Identifiers

2052-49-5
259.48
UN3267
Tetra-n-butylammonium hydroxide
[CH3(CH2)3]4NOH
MFCD00009425
3571228

Specifications

2052-49-5
Liquid
259.48
UN3267
1.435
14
1kg
Tetra-n-butylammonium hydroxide
0.97g/mL
[CH3(CH2)3]4NOH
MFCD00009425
3571228
Colorless to light yellow
55% w/w aq. soln.
Air Sensitive

Safety and Handling

SDS & Safety Documents

GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.

GHS P Statement
P280-P305+P351+P338-P309-P310
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
IF exposed or if you feel unwell: Immediately call a POISON CENTER or doctor/physician.

Danger

DOTInformation : Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: CORROSIVE LIQUID, BASIC, ORGANIC, N.O.S.

RTECSNumber : BS5425000

TSCA : TSCA

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit