Alfa Aesar™ Tetra-n-butylammonium hydroxide, Reagent Grade, 31% in methanol

As a basic nonaqueous titrant

Manufacturer: Alfa Aesar™ 1626314

Quantity 25g

 View more versions of this product

Catalog No. AA1626314

 N/A
N/A

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000. 


    Chemical Identifiers

    CAS 2052-49-5
    Formula Weight 259.48
    MDL Number MFCD00009425
    UN Number UN2924
    Chemical Name or Material Tetra-n-butylammonium hydroxide
    Grade Reagent Grade

    Specifications

    Formula Weight 259.48
    Health Hazard 3 GHS P Statement
    P210-P301+P310-P303+P361+P353-P305+P351+P338-P361-P405-P501a
    Keep away from heat/sparks/open flames/hot surfaces.
    No smoking.
    IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
    IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing.
    Rinse skin with water/shower.
    IF IN EYES: Rinse cautiously with water for several minutes.
    Remove contact lenses, if present and easy to do.
    Continue rinsing.
    Remove/Take off immediately all contaminated clothing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.
    Density 0.830g/mL
    Physical Form Liquid
    Health Hazard 2 GHS H Statement
    H224-H300-H310-H314-H318
    Extremely flammable liquid and vapor.
    Fatal if swallowed.
    Fatal in contact with skin.
    Causes severe skin burns and eye damage.
    Causes serious eye damage.
    Odor Alcohol-like
    CAS 2052-49-5
    Health Hazard 1 Danger
    Refractive Index 1.3775
    Flash Point 13.9° (57°F)
    DOT Information Transport Hazard Class: 3; Packing Group: II; Proper Shipping Name: FLAMMABLE LIQUIDS, CORROSIVE, N.O.S.
    TSCA TSCA
    MDL Number MFCD00009425
    UN Number UN2924
    Concentration 31% in methanol
    Quantity 25g
    Chemical Name or Material Tetra-n-butylammonium hydroxide
    Grade Reagent Grade

    GHS H Statement
    H224-H300-H310-H314-H318
    Extremely flammable liquid and vapor.
    Fatal if swallowed.
    Fatal in contact with skin.
    Causes severe skin burns and eye damage.
    Causes serious eye damage.

    GHS P Statement
    P210-P301+P310-P303+P361+P353-P305+P351+P338-P361-P405-P501a
    Keep away from heat/sparks/open flames/hot surfaces.
    No smoking.
    IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
    IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing.
    Rinse skin with water/shower.
    IF IN EYES: Rinse cautiously with water for several minutes.
    Remove contact lenses, if present and easy to do.
    Continue rinsing.
    Remove/Take off immediately all contaminated clothing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.

    Danger

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit