Alfa Aesar™ Tetra-n-butylammonium hydroxide, Reagent Grade, 31% in methanol

As a basic nonaqueous titrant

Manufacturer: Alfa Aesar™ 16263-22

Quantity 100g

 View more versions of this product

Catalog No. AA1626322

 N/A
N/A

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000. 


    Chemical Identifiers

    CAS 2052-49-5
    Formula Weight 259.48
    MDL Number MFCD00009425
    UN Number UN2924
    Chemical Name or Material Tetra-n-butylammonium hydroxide
    Grade Reagent Grade

    Specifications

    Formula Weight 259.48
    Health Hazard 3 GHS P Statement
    P210-P301+P310-P303+P361+P353-P305+P351+P338-P361-P405-P501a
    Keep away from heat/sparks/open flames/hot surfaces.
    No smoking.
    IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
    IF ON SKIN (or hair): R
    Flash Point 13.9° (57°F)
    Odor Alcohol-like
    Quantity 100g
    DOT Information Transport Hazard Class: 3; Packing Group: II; Proper Shipping Name: FLAMMABLE LIQUIDS, CORROSIVE, N.O.S.
    Density 0.830g/mL
    Physical Form Liquid
    UN Number UN2924
    Health Hazard 1 Danger
    Refractive Index 1.3775
    TSCA TSCA
    MDL Number MFCD00009425
    CAS 2052-49-5
    Health Hazard 2 GHS H Statement
    H224-H300-H310-H314-H318
    Extremely flammable liquid and vapor.
    Fatal if swallowed.
    Fatal in contact with skin.
    Causes severe skin burns and eye damage.
    Causes serious eye damage.
    Concentration 31% in methanol
    Chemical Name or Material Tetra-n-butylammonium hydroxide
    Grade Reagent Grade

    GHS H Statement
    H224-H300-H310-H314-H318
    Extremely flammable liquid and vapor.
    Fatal if swallowed.
    Fatal in contact with skin.
    Causes severe skin burns and eye damage.
    Causes serious eye damage.

    GHS P Statement
    P210-P301+P310-P303+P361+P353-P305+P351+P338-P361-P405-P501a
    Keep away from heat/sparks/open flames/hot surfaces.
    No smoking.
    IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
    IF ON SKIN (or hair): R

    Danger

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit