Alfa Aesar™ Tetra-n-butylammonium phosphate, 98%, non-UV

Manufacturer: Alfa Aesar™ 4674609

Quantity 10g

 View more versions of this product

Catalog No. AA4674609

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 5574-97-0
    Molecular Formula C16H38NO4P
    Linear Formula [CH3(CH2)3]4NH2PO4
    Formula Weight 339.46
    MDL Number MFCD00064526
    Beilstein 5196532
    Synonym Tetra-n-butylammonium dihydrogen phosphate; Tetra-n-butylammonium phosphate monobasic
    Chemical Name or Material Tetra-n-butylammonium phosphate non-UV

    Specifications

    Formula Weight 339.46
    Health Hazard 3 GHS P Statement
    P280-P305+P351+P338-P362-P321-P332+P313-P337+P313
    Wear protective gloves/protective clothing/eye protection/face protection.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
    Continue rinsing.
    Take off contaminated clothing and wash before reuse.
    Specific treatment (see label).
    If skin irritation occurs: Get medical advice/attention.
    If eye irritation persists: Get medical advice/attention.
    Beilstein 5196532
    TSCA TSCA
    Physical Form Crystalline
    Molecular Formula C16H38NO4P
    MDL Number MFCD00064526
    CAS 5574-97-0
    Synonym Tetra-n-butylammonium dihydrogen phosphate; Tetra-n-butylammonium phosphate monobasic
    Health Hazard 1 Warning
    Health Hazard 2 GHS H Statement
    H315-H319
    Causes skin irritation.
    Causes serious eye irritation.
    Color White
    Quantity 10g
    Linear Formula [CH3(CH2)3]4NH2PO4
    Melting Point 151° to 154°C
    Chemical Name or Material Tetra-n-butylammonium phosphate non-UV
    Assay 98%

    GHS H Statement
    H315-H319
    Causes skin irritation.
    Causes serious eye irritation.

    GHS P Statement
    P280-P305+P351+P338-P362-P321-P332+P313-P337+P313
    Wear protective gloves/protective clothing/eye protection/face protection.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
    Continue rinsing.
    Take off contaminated clothing and wash before reuse.
    Specific treatment (see label).
    If skin irritation occurs: Get medical advice/attention.
    If eye irritation persists: Get medical advice/attention.

    Warning

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit