Learn More
Tetra-n-butylammonium phosphate, 98%, non-UV, Thermo Scientific Chemicals
Description
Tetra-n-butylammonium phosphate, is used in chemicals, pharmaceutical research, pharmaceutical intermediates, and reagents. Tetrabutylammonium phosphate can be used in organic synthesis. It is used as an Ion-associating reagent for HPLC.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Melting Point | 151°C to 154°C |
Color | White |
Linear Formula | [CH3(CH2)3]4NH2PO4 |
Quantity | 50 g |
Beilstein | 5196532 |
Solubility Information | It is soluble in water. |
Formula Weight | 339.46 |
Percent Purity | 98% |
Physical Form | Crystalline |
Chemical Name or Material | Tetra-n-butylammonium phosphate non-UV |
Safety and Handling
Hazard Category | H500 |
Hazard Statement | GHS H Statement H315-H319 Causes skin irritation. Causes serious eye irritation. |
Precautionary Statement | GHS P Statement P280-P305+P351+P338-P362-P321-P332+P313-P337+P313 Wear protective gloves/protective clothing/eye protection/face protection. IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing. Take off contaminated clothing and wash before reuse. Specific treatment (see label). If skin irritation occurs: Get medical advice/attention. If eye irritation persists: Get medical advice/attention. |
EINECSNumber | 226-947-1 |
TSCA | Yes |
Recommended Storage | Ambient temperatures |
RUO – Research Use Only
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.