Alfa Aesar™ Tetra-n-butylammonium tetra-n-butylborate, 97%

Manufacturer: Alfa Aesar™ H5551206

Quantity 5g

 View more versions of this product

Catalog No. AAH5551206

 N/A
N/A

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000. 


    Chemical Identifiers

    CAS 23231-91-6
    Linear Formula [CH3(CH2)3]4N(B[(CH2)3CH3]4)
    Formula Weight 481.74
    MDL Number MFCD00061526
    Beilstein 4171555
    Chemical Name or Material Tetra-n-butylammonium tetra-n-butylborate

    Specifications

    Formula Weight 481.74
    Melting Point 106° to 109°C
    Physical Form Powder
    MDL Number MFCD00061526
    Beilstein 4171555
    Color White
    Linear Formula [CH3(CH2)3]4N(B[(CH2)3CH3]4)
    CAS 23231-91-6
    Quantity 5g
    Chemical Name or Material Tetra-n-butylammonium tetra-n-butylborate
    Assay 97%

  • Description & Specifications

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit