Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ Tetra-n-butylammonium tetra-n-butylborate, 97%

Manufacturer:  Alfa Aesar™ H5551203

Quantity 1g

 View more versions of this product

Catalog No. AAH5551203

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Specifications Description & Specifications

Chemical Identifiers

23231-91-6
481.74
4171555
[CH3(CH2)3]4N(B[(CH2)3CH3]4)
MFCD00061526
Tetra-n-butylammonium tetra-n-butylborate

Specifications

White
1g
Powder
481.74
4171555
97%
106° to 109°C
23231-91-6
[CH3(CH2)3]4N(B[(CH2)3CH3]4)
MFCD00061526
Tetra-n-butylammonium tetra-n-butylborate
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit