Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tetra-n-butylammonium tribromide, 98+%, ACROS Organics™

Click to view available options
Quantity:
25g
100g
500g
Packaging:
Glass bottle
38932-80-8
C16H36Br3N
482.15
SFLXUZPXEWWQNH-UHFFFAOYSA-K
tetrabutylammonium tribromide,tetra-n-butylammonium tribromide,tetrabutylammoniumtribromide,mono tetrabutylammonium tribromide,tetrabutylamoniumtribromide,acmc-1ct1n,tetrabutylamonium tribromide,tetrabutylazanium tribromide,tetrabutylammonium-tribromide,tetra-butylammonium tribromide
23500184
tetrabutylazanium;tribromide
CCCC[N+](CCCC)(CCCC)CCCC.CCCC[N+](CCCC)(CCCC)CCCC.CCCC[N+](CCCC)(CCCC)CCCC.[Br-].[Br-].[Br-]
38932-80-8
C16H36Br3N
482.15
SFLXUZPXEWWQNH-UHFFFAOYSA-K
tetrabutylammonium tribromide,tetra-n-butylammonium tribromide,tetrabutylammoniumtribromide,mono tetrabutylammonium tribromide,tetrabutylamoniumtribromide,acmc-1ct1n,tetrabutylamonium tribromide,tetrabutylazanium tribromide,tetrabutylammonium-tribromide,tetra-butylammonium tribromide
23500184
tetrabutylazanium;tribromide
CCCC[N+](CCCC)(CCCC)CCCC.CCCC[N+](CCCC)(CCCC)CCCC.CCCC[N+](CCCC)(CCCC)CCCC.[Br-].[Br-].[Br-]
C-X bond formation reagent
Melting Point 71.0°C to 75.0°C
Quantity 25g
Packaging Glass bottle
Infrared Spectrum Authentic
Color Orange
Assay Percent Range 98+%
Linear Formula [CH3(CH2)3]4NBr3
Solubility Information Solubility in water: insoluble in water. Other solubilities: slightly soluble in alcohols, well soluble in acetonitrille,chcl3,thf,dioxane
Formula Weight 482.15
Physical Form Crystalline Powder
Percent Purity ≥98%
Chemical Name or Material Tetra-n-butylammonium tribromide
Show More Show Less