Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ Tetra-n-butylgermanium, 99%

Click to view available options
Quantity:
1g
5g
25g
1067-42-1
C16H36Ge
301.094
MFCD00015224
HDVLQIDIYKIVRE-UHFFFAOYSA-N
tetrabutylgermanium,germane, tetrabutyl,tetra-n-butylgermane,tetra-n-butylgermanium,tetrabutyl-german,tetrabuthylgermanium,tetrabutyl germanium
70599
tetrabutylgermane
CCCC[Ge](CCCC)(CCCC)CCCC
1067-42-1
C16H36Ge
301.094
MFCD00015224
HDVLQIDIYKIVRE-UHFFFAOYSA-N
tetrabutylgermanium,germane, tetrabutyl,tetra-n-butylgermane,tetra-n-butylgermanium,tetrabutyl-german,tetrabuthylgermanium,tetrabutyl germanium
70599
tetrabutylgermane
CCCC[Ge](CCCC)(CCCC)CCCC
Refractive Index 1.4571
Flash Point >110°C (230°F)
Density 0.934 g/mL at 20°C
Quantity 25g
Boiling Point 277°C to 279°C
Formula Weight 301.05
Physical Form Liquid
Percent Purity 99%
Chemical Name or Material Tetra-n-butylgermanium

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P264b-P270-P301+P312-P330-P501c

H302

EINECSNumber : 213-929-3

TSCA : Yes

Recommended Storage : Ambient temperatures