Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ Tetra-n-butyllead, 95%

Manufacturer:  Alfa Aesar™ 5712903

Quantity 1g

 View more versions of this product

Catalog No. AA5712903


  Check Availability
Add to cart

Specifications Description & Specifications

Chemical Identifiers

1920-90-7
[CH3(CH2)3]4Pb
MFCD00015226
Lead tetrabutyl; Tetra-n-butylplumbane
C16H36Pb
435.66
UN3282
Tetra-n-butyllead

Specifications

1g
1.324g/mL
Colorless to yellow
Air & Light Sensitive
Liquid, May Include Precipitate
[CH3(CH2)3]4Pb
MFCD00015226
Lead tetrabutyl; Tetra-n-butylplumbane
Tetra-n-butyllead
1920-90-7
140°C (1mmHg)
140°C (284°F)
1.5119
C16H36Pb
435.66
UN3282
Insoluble in water
95%

Safety and Handling

SDS & Safety Documents

GHS H Statement
H300-H310-H330-H360-H373
Fatal if swallowed.
Fatal in contact with skin.
Fatal if inhaled.
May damage fertility or the unborn child.
May cause damage to organs through prolonged or repeated exposure.

GHS P Statement
P260-P301+P310-P304+P340-P320-P330-P361-P405-P501a
Do not breathe dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Specific treatment is urgent (see label).
Rinse mouth.
Remove/Take off immediately all contaminated clothing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Danger

DOTInformation : Transport Hazard Class: 6.1; Packing Group: II; Proper Shipping Name: ORGANOMETALLIC COMPOUND, TOXIC, LIQUID, N.O.S.

TSCA : TSCA

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit