Alfa Aesar™ Tetra-n-butylphosphonium bromide, 99%

Manufacturer: Alfa Aesar™ A1086814

Quantity 25g

 View more versions of this product

Catalog No. AAA1086814

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 3115-68-2
    Molecular Formula C16H36BrP
    Linear Formula [CH3(CH2)3]4PBr
    Formula Weight 339.35
    MDL Number MFCD00011853
    UN Number UN3464
    Beilstein 4160474
    Chemical Name or Material Tetra-n-butylphosphonium bromide

    Specifications

    Formula Weight 339.35
    Health Hazard 3 GHS P Statement
    P261-P305+P351+P338-P361-P302+P350-P405-P501a
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
    Continue rinsing.
    Remove/Take off immediately all contaminated clothing.
    IF ON SKIN: Gently wash with plenty of soap and water.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.
    TSCA TSCA
    Linear Formula [CH3(CH2)3]4PBr
    Flash Point 290°C (554°F)
    Beilstein 4160474
    Health Hazard 1 Danger
    Health Hazard 2 GHS H Statement
    H310-H318-H302-H332-H315-H335
    Fatal in contact with skin.
    Causes serious eye damage.
    Harmful if swallowed.
    Harmful if inhaled.
    Causes skin irritation.
    May cause respiratory irritation.
    Melting Point 99° to 103°C
    RTECS Number TA2417000
    CAS 3115-68-2
    UN Number UN3464
    Quantity 25g
    Molecular Formula C16H36BrP
    MDL Number MFCD00011853
    DOT Information Transport Hazard Class: 6.1; Packing Group: II; Proper Shipping Name: ORGANOPHOSPHORUS COMPOUND, TOXIC, SOLID, N.O.S.
    Chemical Name or Material Tetra-n-butylphosphonium bromide
    Assay 99%

    Description

    Applications
    Tetra-n-butylphosphonium bromide is used as a phase-transfer catalyst at high temperatures, at which nitrogen analogues tend to undergo elimination e.g. as catalyst for the dealkoxycarbonylation of malonates and ß-keto esters by heating in stearic acid. used as an ionic liquid solvent in the regioselective O-alkylation of C/O ambident nucleophiles. It is a synthetic reagent used in proteomics research.

    Solubility
    Soluble in water (70 g/100 ml)

    Notes
    Store in cool place. Keep container tightly closed in a dry and well-ventilated place. It is hygroscopic in nature.

    GHS H Statement
    H310-H318-H302-H332-H315-H335
    Fatal in contact with skin.
    Causes serious eye damage.
    Harmful if swallowed.
    Harmful if inhaled.
    Causes skin irritation.
    May cause respiratory irritation.

    GHS P Statement
    P261-P305+P351+P338-P361-P302+P350-P405-P501a
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
    Continue rinsing.
    Remove/Take off immediately all contaminated clothing.
    IF ON SKIN: Gently wash with plenty of soap and water.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.

    Danger

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit