Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tetra-n-butyltin, 95%, Thermo Scientific Chemicals

Catalog Number aa1411522
Click to view available options
Quantity:
100 g
500 g
1461-25-2
C16H36Sn
347.174
MFCD00009422
AFCAKJKUYFLYFK-UHFFFAOYSA-N
tetrabutyltin, tetra-n-butyltin, stannane, tetrabutyl, tin, tetrabutyl, tetra-n-butylcin, tetra-n-butylcin czech, unii-qj7y5v377v, ccris 6322, tetra-normal-butyltin, wln: 4-sn-4&4&4
15098
tetrabutylstannane
CCCC[Sn](CCCC)(CCCC)CCCC
1461-25-2
C16H36Sn
347.174
MFCD00009422
AFCAKJKUYFLYFK-UHFFFAOYSA-N
tetrabutyltin, tetra-n-butyltin, stannane, tetrabutyl, tin, tetrabutyl, tetra-n-butylcin, tetra-n-butylcin czech, unii-qj7y5v377v, ccris 6322, tetra-normal-butyltin, wln: 4-sn-4&4&4
15098
tetrabutylstannane
CCCC[Sn](CCCC)(CCCC)CCCC

Tetra-n-butyltin is used in hydrogenolysis reactions and as a transition metal catalyst. It is also used stabilizers in the manufacture of polyvinyl chloride.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Melting Point -97°C
Density 1.057 g/mL
Boiling Point 144°C to 146°C (10 mmHg)
Flash Point 107°C (225°F)
Odor Mild
Refractive Index 1.474
Quantity 100 g
UN Number UN2788
Solubility Information Insoluble in water. It is soluble in most common organic solvents.
Formula Weight 347.17
Percent Purity 95%
Physical Form Liquid
Chemical Name or Material Tetra-n-butyltin
Show More Show Less
Hazard Category H301-H312-H315-H319-H372
Hazard Statement GHS H Statement
H301-H372-H312-H315-H319
Toxic if swallowed.
Causes damage to organs through prolonged or repeated exposure.
Harmful in contact with skin.
Causes skin irritation.
Causes serious eye irritation.
Precautionary Statement P260-P264b-P270-P280-P301+P310-P302+P352-P305+P351+P338-P312-P314-P330-P332+P313-P362-P501c
DOTInformation Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: ORGANOTIN COMPOUNDS, LIQUID, N.O.S.
EINECSNumber 215-960-8
RTECSNumber WH8605000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit