Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tetra-n-butyltin, 96%, ACROS Organics™

Click to view available options
Quantity:
5g
100g
500g
Packaging:
Glass bottle
1461-25-2
C16H36Sn
347.174
MFCD00009422
AFCAKJKUYFLYFK-UHFFFAOYSA-N
tetrabutyltin,tetra-n-butyltin,stannane, tetrabutyl,tin, tetrabutyl,tetra-n-butylcin,tetra-n-butylcin czech,unii-qj7y5v377v,ccris 6322,tetra-normal-butyltin,wln: 4-sn-4&4&4
15098
tetrabutylstannane
CCCC[Sn](CCCC)(CCCC)CCCC
1461-25-2
C16H36Sn
347.174
MFCD00009422
AFCAKJKUYFLYFK-UHFFFAOYSA-N
tetrabutyltin,tetra-n-butyltin,stannane, tetrabutyl,tin, tetrabutyl,tetra-n-butylcin,tetra-n-butylcin czech,unii-qj7y5v377v,ccris 6322,tetra-normal-butyltin,wln: 4-sn-4&4&4
15098
tetrabutylstannane
CCCC[Sn](CCCC)(CCCC)CCCC
Color Colorless to Straw
Quantity 100g
Packaging Glass bottle
Infrared Spectrum Authentic
Density 1.057g/mL
Assay Percent Range 96%
Linear Formula [CH3(CH2)3]4Sn
Formula Weight 347.14
Physical Form Liquid
Percent Purity ≥95%
Chemical Name or Material Tetra-n-butyltin
Show More Show Less

GHS H Statement:
Toxic if swallowed.
Harmful in contact with skin.
Causes skin irritation.
Causes serious eye irritation.
Causes damage to organs through prolonged or repeated exposure.
Very toxic to aquatic life with long lasting effects.

GHS P Statement:
Do not breathe dust/fume/gas/mist/vapors/spray.
Avoid release to the environment.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF SWALLOWED: Immediately call a POISON CENTRE or doctor/physician.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Danger