Alfa Aesar™ Tetra-n-propylammonium hydroxide, 40% w/w aq. soln.

Manufacturer: Alfa Aesar™ 1745606

Quantity 5g

 View more versions of this product

Catalog No. AA1745606

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 4499-86-9
    Linear Formula (CH3CH2CH2)4NOH
    Formula Weight 203.37
    MDL Number MFCD00009360
    UN Number UN3267
    Beilstein 4207983
    Synonym TPAH
    Chemical Name or Material Tetra-n-propylammonium hydroxide

    Specifications

    Formula Weight 203.37
    Health Hazard 3 GHS P Statement
    P260-P303+P361+P353-P305+P351+P338-P301+P330+P331-P405-P501a
    Do not breathe dust/fume/gas/mist/vapors/spray.
    IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing.
    Rinse skin with water/shower.
    IF IN EYES: Rinse cautiously with water for several minutes.
    Remove contact lenses, if present and easy to do.
    Continue rinsing.
    IF SWALLOWED: Rinse mouth.
    Do NOT induce vomiting.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.
    Density 0.99g/mL
    Physical Form Liquid
    CAS 4499-86-9
    Synonym TPAH
    Health Hazard 1 Danger
    Refractive Index 1.3716
    Health Hazard 2 GHS H Statement
    H314
    Causes severe skin burns and eye damage.
    DOT Information Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: CORROSIVE LIQUID, BASIC, ORGANIC, N.O.S.
    RTECS Number BS8400000
    TSCA TSCA
    Linear Formula (CH3CH2CH2)4NOH
    MDL Number MFCD00009360
    Beilstein 4207983
    Boiling Point 102°C
    Color Colorless to pale yellow
    Sensitivity Air Sensitive
    UN Number UN3267
    Concentration 40% w/w aq. soln.
    Quantity 5g
    Chemical Name or Material Tetra-n-propylammonium hydroxide

    GHS H Statement
    H314
    Causes severe skin burns and eye damage.

    GHS P Statement
    P260-P303+P361+P353-P305+P351+P338-P301+P330+P331-P405-P501a
    Do not breathe dust/fume/gas/mist/vapors/spray.
    IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing.
    Rinse skin with water/shower.
    IF IN EYES: Rinse cautiously with water for several minutes.
    Remove contact lenses, if present and easy to do.
    Continue rinsing.
    IF SWALLOWED: Rinse mouth.
    Do NOT induce vomiting.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.

    Danger

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit