Alfa Aesar™ Tetra-n-propylammonium perruthenate(VII), 98%

Manufacturer: Alfa Aesar™ B2451177

Quantity 0.1g

 View more versions of this product

Catalog No. AAB2451177

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 114615-82-6
    Molecular Formula C12H28NO4Ru
    Linear Formula (CH3(CH2)2)4NRuO4
    Formula Weight 351.43
    MDL Number MFCD00074914
    UN Number UN1479
    Synonym TPAP
    Chemical Name or Material Tetra-n-propylammonium perruthenate(VII)

    Specifications

    Formula Weight 351.43
    Molecular Formula C12H28NO4Ru
    Linear Formula (CH3(CH2)2)4NRuO4
    MDL Number MFCD00074914
    CAS 114615-82-6
    UN Number UN1479
    Synonym TPAP
    Melting Point 160°C (dec.)
    Quantity 0.1g
    Health Hazard 1 Danger
    Health Hazard 2 GHS H Statement
    H272-H315-H319-H335
    May intensify fire; oxidizer.
    Causes skin irritation.
    Causes serious eye irritation.
    May cause respiratory irritation.
    Health Hazard 3 GHS P Statement
    P221-P210-P220-P305+P351+P338-P405-P501a
    Take any precaution to avoid mixing with combustibles.
    Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
    Keep/Store away from clothing/combustible materials.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.
    Recommended Storage 2° to 8°C
    DOT Information Transport Hazard Class: 5.1; Packing Group: III; Proper Shipping Name: OXIDIZING SOLID, N.O.S.
    Chemical Name or Material Tetra-n-propylammonium perruthenate(VII)
    Assay 98%

    Description

    Applications
    Tetra-n-propylammonium perruthenate a catalytic oxidant used in conjunction with N-methylmorpholine N-oxide for oxidation of primary & secondary alcohols to aldehydes and ketones.

    Solubility
    Insoluble in water.

    Notes
    Hygroscopic. Store under inert gas. Store away from oxidizing agents, reducing agents, heat, organic materials, metal powders, heat.

    GHS H Statement
    H272-H315-H319-H335
    May intensify fire; oxidizer.
    Causes skin irritation.
    Causes serious eye irritation.
    May cause respiratory irritation.

    2° to 8°C

    GHS P Statement
    P221-P210-P220-P305+P351+P338-P405-P501a
    Take any precaution to avoid mixing with combustibles.
    Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
    Keep/Store away from clothing/combustible materials.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.

    Danger

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit