Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tetra-n-propylammonium perruthenate(VII), 98%, Thermo Scientific Chemicals

Catalog Number AAB2451102
Click to view available options
Quantity:
0.5 g
114615-82-6
C12H35NO4Ru
358.485
MFCD00074914
HUCLFLGLPCVDMZ-UHFFFAOYSA-M
1-propanaminium, n,n,n-tripropyl-, t-4-tetraoxoruthenate 1-
127020979
ruthenium;tetrapropylazanium;hydroxide;trihydrate
CCC[N+](CCC)(CCC)CCC.O.O.O.[OH-].[Ru]
114615-82-6
C12H35NO4Ru
358.485
MFCD00074914
HUCLFLGLPCVDMZ-UHFFFAOYSA-M
1-propanaminium, n,n,n-tripropyl-, t-4-tetraoxoruthenate 1-
127020979
ruthenium;tetrapropylazanium;hydroxide;trihydrate
CCC[N+](CCC)(CCC)CCC.O.O.O.[OH-].[Ru]

Tetra-n-propylammonium perruthenate a catalytic oxidant used in conjunction with N-methylmorpholine N-oxide for oxidation of primary & secondary alcohols to aldehydes and ketones.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Tetra-n-propylammonium perruthenate a catalytic oxidant used in conjunction with N-methylmorpholine N-oxide for oxidation of primary & secondary alcohols to aldehydes and ketones.

Solubility
Insoluble in water.

Notes
Hygroscopic. Store under inert gas. Store away from oxidizing agents, reducing agents, heat, organic materials, metal powders, heat.
TRUSTED_SUSTAINABILITY
Melting Point 160°C (decomposition)
Quantity 0.5 g
Linear Formula (CH3(CH2)2)4NRuO4
UN Number UN1479
Solubility Information Insoluble in water.
Formula Weight 351.43
Percent Purity 98%
Sensitivity Hygroscopic
Chemical Name or Material Tetra-n-propylammonium perruthenate(VII)
Hazard Category H272-H315-H319-H335-H500
Hazard Statement GHS H Statement
H272-H315-H319-H335
May intensify fire; oxidizer.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P210-P220-P221-P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P370+P378q-P501c
DOTInformation Hazard Class: 5.1; Packaging Group: II
TSCA No
Recommended Storage Keep cold; Store under Nitrogen

RUO – Research Use Only

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit