Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tetraacetylethylenediamine, 90%, ACROS Organics™

Click to view available options
Quantity:
50g
250g
Packaging:
Plastic bottle
10543-57-4
C10H16N2O4
228.25
MFCD00014967
BGRWYDHXPHLNKA-UHFFFAOYSA-N
tetraacetylethylenediamine,n,n,n',n'-tetraacetylethylenediamine,taed,tetracetylethylenediamine,acetamide, n,n'-1,2-ethanediylbis n-acetyl,n,n'-ethylenebis n-acetylacetamide,unii-p411ed0n2b,n,n'-ethane-1,2-diyl bis n-acetylacetamide,n-acetyl-n-2-diacetylamino ethyl acetamide
66347
N-acetyl-N-[2-(diacetylamino)ethyl]acetamide
CC(=O)N(CCN(C(=O)C)C(=O)C)C(=O)C
10543-57-4
C10H16N2O4
228.25
MFCD00014967
BGRWYDHXPHLNKA-UHFFFAOYSA-N
tetraacetylethylenediamine,n,n,n',n'-tetraacetylethylenediamine,taed,tetracetylethylenediamine,acetamide, n,n'-1,2-ethanediylbis n-acetyl,n,n'-ethylenebis n-acetylacetamide,unii-p411ed0n2b,n,n'-ethane-1,2-diyl bis n-acetylacetamide,n-acetyl-n-2-diacetylamino ethyl acetamide
66347
N-acetyl-N-[2-(diacetylamino)ethyl]acetamide
CC(=O)N(CCN(C(=O)C)C(=O)C)C(=O)C
Melting Point 149°C to 154°C
Flash Point 140°C
Boiling Point 140°C (2.0 hPa)
Infrared Spectrum Authentic
Color White to Beige
Quantity 250g
Packaging Plastic bottle
Assay Percent Range 88% min
Merck Index 15, 9158
Solubility Information Solubility in water: ca 1g/L (20°C). Other solubilities: slightly soluble in acetone,moderately soluble in chloroform,methylenechloride
Formula Weight 228.25
Physical Form Granular Powder
Percent Purity 90%
Chemical Name or Material Tetraacetylethylenediamine, 90%
Show More Show Less

EINECSNumber : 234-123-8

TSCA : TSCA