Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ Tetraallyltin, 96%

Click to view available options
Quantity:
2g
10g
7393-43-3
C12H20Sn
283.002
MFCD00008637
XJPKDRJZNZMJQM-UHFFFAOYSA-N
tetraallyltin,tetraallylstannane,tetrallylstannane,stannane, tetra-2-propenyl,stannane, tetraallyl,acmc-209osy,tetrakis prop-2-enyl stannane,stannane,tetra-2-propen-1-yl,tetrakis prop-2-en-1-yl stannane
81878
tetrakis(prop-2-enyl)stannane
C=CC[Sn](CC=C)(CC=C)CC=C
7393-43-3
C12H20Sn
283.002
MFCD00008637
XJPKDRJZNZMJQM-UHFFFAOYSA-N
tetraallyltin,tetraallylstannane,tetrallylstannane,stannane, tetra-2-propenyl,stannane, tetraallyl,acmc-209osy,tetrakis prop-2-enyl stannane,stannane,tetra-2-propen-1-yl,tetrakis prop-2-en-1-yl stannane
81878
tetrakis(prop-2-enyl)stannane
C=CC[Sn](CC=C)(CC=C)CC=C
Refractive Index 1.5385
Quantity 2g
Boiling Point 69°C to 70°C (1.5mmHg)
Density 1.179 g/mL
Sensitivity Air Sensitive
Flash Point 75°C (167°F)
UN Number UN2788
Solubility Information Not miscible in water or difficult to mix.
Formula Weight 282.98
Physical Form Liquid
Percent Purity 96%
Chemical Name or Material Tetraallyltin
Show More Show Less

GHS H Statement
H300-H310-H330-H227-H315-H319
Fatal if swallowed.
Fatal in contact with skin.
Fatal if inhaled.
Combustible liquid.
Causes skin irritation.
Causes serious eye irritation.

P210-P235-P261-P264b-P270-P271-P280-P301+P310-P302+P352-P304+P340-P311-P312-P330-P361-P363-P370+P378q-P501c

H227-H301+H311+H331-H335

DOTInformation : Transport Hazard Class: 6.1; Packing Group: II; Proper Shipping Name: ORGANOTIN COMPOUNDS, LIQUID, N.O.S.

EINECSNumber : 230-987-5

TSCA : No

Recommended Storage : Ambient temperatures