Alfa Aesar™ Tetraallyltin, 96%

Manufacturer: Alfa Aesar™ 7115804

Quantity 2g

 View more versions of this product

Catalog No. AA7115804

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 7393-43-3
    Molecular Formula Sn(CH2CH=CH2)4
    Formula Weight 282.98
    MDL Number MFCD00008637
    UN Number UN2788
    Chemical Name or Material Tetraallyltin

    Specifications

    Formula Weight 282.98
    Health Hazard 3 GHS P Statement
    P210-P301+P310-P304+P340-P305+P351+P338-P320-P330-P361-P405-P501a
    Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
    IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
    F INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
    Specific treatment is urgent (see ... on this label).
    Rinse mouth.
    Remove/Take off immediately all contaminated clothing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.
    Physical Form Liquid
    Molecular Formula Sn(CH2CH=CH2)4
    CAS 7393-43-3
    DOT Information Transport Hazard Class: 6.1; Packing Group: II; Proper Shipping Name: ORGANOTIN COMPOUNDS, LIQUID, N.O.S.
    Sensitivity Air Sensitive
    Boiling Point 69° to 70°C (1.5mmHg)
    MDL Number MFCD00008637
    Quantity 2g
    UN Number UN2788
    Flash Point 75°C (167°F)
    Health Hazard 1 Danger
    Health Hazard 2 GHS H Statement
    H300-H310-H330-H227-H315-H319
    Fatal if swallowed.
    Fatal in contact with skin.
    Fatal if inhaled.
    Combustible liquid.
    Causes skin irritation.
    Causes serious eye irritation.
    Density 1.179g/mL
    Refractive Index 1.5385
    Chemical Name or Material Tetraallyltin
    Assay 96%

    GHS H Statement
    H300-H310-H330-H227-H315-H319
    Fatal if swallowed.
    Fatal in contact with skin.
    Fatal if inhaled.
    Combustible liquid.
    Causes skin irritation.
    Causes serious eye irritation.

    GHS P Statement
    P210-P301+P310-P304+P340-P305+P351+P338-P320-P330-P361-P405-P501a
    Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
    IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
    F INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
    Specific treatment is urgent (see ... on this label).
    Rinse mouth.
    Remove/Take off immediately all contaminated clothing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.

    Danger

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit