Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tetraallyltin, 97%, ACROS Organics™

Click to view available options
Quantity:
1g
Packaging:
Glass Bottle
7393-43-3
C12H20Sn
283.002
MFCD00008637
XJPKDRJZNZMJQM-UHFFFAOYSA-N
tetraallyltin,tetraallylstannane,tetrallylstannane,stannane, tetra-2-propenyl,stannane, tetraallyl,acmc-209osy,tetrakis prop-2-enyl stannane,stannane,tetra-2-propen-1-yl,tetrakis prop-2-en-1-yl stannane
81878
tetrakis(prop-2-enyl)stannane
C=CC[Sn](CC=C)(CC=C)CC=C
7393-43-3
C12H20Sn
283.002
MFCD00008637
XJPKDRJZNZMJQM-UHFFFAOYSA-N
tetraallyltin,tetraallylstannane,tetrallylstannane,stannane, tetra-2-propenyl,stannane, tetraallyl,acmc-209osy,tetrakis prop-2-enyl stannane,stannane,tetra-2-propen-1-yl,tetrakis prop-2-en-1-yl stannane
81878
tetrakis(prop-2-enyl)stannane
C=CC[Sn](CC=C)(CC=C)CC=C
Packaging Glass Bottle
Flash Point 75°C
Density 1.1790g/mL
Color Colorless
Refractive Index 1.5400 to 1.5420 (20°C, 589nm)
Infrared Spectrum Authentic
Specific Gravity 1.179
Boiling Point 69.0°C to 70.0°C (1.5mmHg)
Quantity 1g
Assay Percent Range 97%
Linear Formula (CH2=CHCH2)4Sn
Formula Weight 282.98
Physical Form Liquid
Percent Purity 97%
Chemical Name or Material Tetraallyltin
Show More Show Less

GHS H Statement:
Toxic if swallowed.
Toxic in contact with skin.
Toxic if inhaled.

GHS P Statement:
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
Wear protective gloves/protective clothing/eye protection/face protection.
Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Gently wash with plenty of soap and water.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

Danger