Alfa Aesar™ Tetraamminepalladium(II) chloride monohydrate, 99.9% (metals basis), Pd 39% min

Manufacturer: Alfa Aesar™ 1103609

Quantity 10g

 View more versions of this product

Catalog No. AA1103609

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 13933-31-8
    Formula Weight 263.45 (245.43 Anhydrous)
    MDL Number MFCD00151033
    UN Number UN3077
    Chemical Name or Material Tetraamminepalladium(II) chloride monohydrate

    Specifications

    Formula Weight 263.45 (245.43 Anhydrous)
    Health Hazard 3 GHS P Statement
    P261-P280-P305+P351+P338-P301+P312-P406-P501a
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    Wear protective gloves/protective clothing/eye protection/face protection.
    IF IN EYES: Rinse cautiously with water for several minutes.
    Remove contact lenses, if present and easy to do.
    Continue rinsing.
    IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
    Store in corrosive resistant container with a resistant inner liner.
    Dispose of contents/container in accordance with local/regional/national/international regulations.
    MDL Number MFCD00151033
    TSCA TSCA
    Physical Form Crystalline
    UN Number UN3077
    Color Colorless to yellow
    DOT Information Transport Hazard Class: 9; Packing Group: III; Proper Shipping Name: ENVIRONMENTALLY HAZARDOUS SUBSTANCES, SOLID, N.O.S.
    Density 1.91g/mL
    Health Hazard 1 Warning
    Concentration Pd 39% min.
    Quantity 10g
    Sensitivity Hygroscopic
    CAS 13933-31-8
    Health Hazard 2 GHS H Statement
    H290-H302-H317-H319
    May be corrosive to metals.
    Harmful if swallowed.
    May cause an allergic skin reaction.
    Causes serious eye irritation.
    Melting Point 120°C (dec.)
    Chemical Name or Material Tetraamminepalladium(II) chloride monohydrate
    Assay 99.9%, (metals basis)

    GHS H Statement
    H290-H302-H317-H319
    May be corrosive to metals.
    Harmful if swallowed.
    May cause an allergic skin reaction.
    Causes serious eye irritation.

    GHS P Statement
    P261-P280-P305+P351+P338-P301+P312-P406-P501a
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    Wear protective gloves/protective clothing/eye protection/face protection.
    IF IN EYES: Rinse cautiously with water for several minutes.
    Remove contact lenses, if present and easy to do.
    Continue rinsing.
    IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
    Store in corrosive resistant container with a resistant inner liner.
    Dispose of contents/container in accordance with local/regional/national/international regulations.

    Warning

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit