Alfa Aesar™ Tetraamminepalladium(II) hydrogen carbonate

Manufacturer: Alfa Aesar™ 4508214

Quantity 25g

 View more versions of this product

Catalog No. AA4508214

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 134620-00-1
    Molecular Formula C2H14N4O6Pd
    Linear Formula [Pd(NH3)4](HCO3)2
    Formula Weight 296.58
    MDL Number MFCD09953449
    UN Number UN3077
    Chemical Name or Material Tetraamminepalladium (II) hydrogen carbonate

    Specifications

    Formula Weight 296.58
    Health Hazard 3 GHS P Statement
    P261-P280-P273-P305+P351+P338-P309+P311-P501a
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    Wear protective gloves/protective clothing/eye protection/face protection.
    Avoid release to the environment.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
    Continue rinsing.
    IF exposed or if you feel unwell: Call a POISON CENTER or doctor/physician.
    Dispose of contents/container in accordance with local/regional/national/international regulations.
    Molecular Formula C2H14N4O6Pd
    Density 2.04g/mL
    CAS 134620-00-1
    Color Colorless to light yellow
    Quantity 25g
    DOT Information Transport Hazard Class: 9; Packing Group: III; Proper Shipping Name: ENVIRONMENTALLY HAZARDOUS SUBSTANCES, SOLID, N.O.S.
    TSCA TSCA
    Linear Formula [Pd(NH3)4](HCO3)2
    UN Number UN3077
    Health Hazard 2 GHS H Statement
    H318-H371-H373-H302-H317
    Causes serious eye damage.
    May cause damage to organs.
    May cause damage to organs through prolonged or repeated exposure.
    Harmful if swallowed.
    May cause an allergic skin reaction.
    RTECS Number RT3597200
    Physical Form Crystalline
    MDL Number MFCD09953449
    Health Hazard 1 Danger
    Chemical Name or Material Tetraamminepalladium (II) hydrogen carbonate

    GHS H Statement
    H318-H371-H373-H302-H317
    Causes serious eye damage.
    May cause damage to organs.
    May cause damage to organs through prolonged or repeated exposure.
    Harmful if swallowed.
    May cause an allergic skin reaction.

    GHS P Statement
    P261-P280-P273-P305+P351+P338-P309+P311-P501a
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    Wear protective gloves/protective clothing/eye protection/face protection.
    Avoid release to the environment.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
    Continue rinsing.
    IF exposed or if you feel unwell: Call a POISON CENTER or doctor/physician.
    Dispose of contents/container in accordance with local/regional/national/international regulations.

    Danger

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit