Alfa Aesar™ Tetraamminepalladium(II) nitrate solution, 99.9% (metals basis), Pd 5.0% max (cont. Pd)

Manufacturer: Alfa Aesar™ 35749NC

Quantity 10g

 View more versions of this product

Catalog No. AA35749NC

 N/A
N/A

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000. 


    Chemical Identifiers

    CAS 13601-08-6
    Formula Weight 298.63
    MDL Number MFCD00011591
    UN Number UN3218
    Chemical Name or Material Tetraamminepalladium(II) nitrate solution

    Specifications

    Formula Weight 298.63
    Health Hazard 3 GHS P Statement
    P221-P210-P220-P261-P305+P351+P338-P501a
    Take any precaution to avoid mixing with combustibles.
    Keep away from heat/sparks/open flames/hot surfaces.
    No smoking.
    Keep/Store away from clothing/combustible materials.
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    IF IN EYES: Rinse cautiously with water for several minutes.
    Remove contact lenses, if present and easy to do.
    Continue rinsing.
    Dispose of contents/container in accordance with local/regional/national/international regulations.
    Physical Form Liquid
    MDL Number MFCD00011591
    CAS 13601-08-6
    Color Colorless to pale yellow
    TSCA TSCA
    UN Number UN3218
    Health Hazard 1 Danger
    Quantity 10g
    DOT Information Transport Hazard Class: 5.1; Packing Group: III; Proper Shipping Name: NITRATES, INORGANIC, AQUEOUS SOLUTION, N.O.S .
    Health Hazard 2 GHS H Statement
    H272-H302-H319-H317
    May intensify fire
    oxidizer.
    Harmful if swallowed.
    Causes serious eye irritation.
    May cause an allergic skin reaction.
    Concentration Pd 5.0% max. (cont. Pd)
    Chemical Name or Material Tetraamminepalladium(II) nitrate solution
    Assay 99.9%, (metals basis)

    GHS H Statement
    H272-H302-H319-H317
    May intensify fire
    oxidizer.
    Harmful if swallowed.
    Causes serious eye irritation.
    May cause an allergic skin reaction.

    GHS P Statement
    P221-P210-P220-P261-P305+P351+P338-P501a
    Take any precaution to avoid mixing with combustibles.
    Keep away from heat/sparks/open flames/hot surfaces.
    No smoking.
    Keep/Store away from clothing/combustible materials.
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    IF IN EYES: Rinse cautiously with water for several minutes.
    Remove contact lenses, if present and easy to do.
    Continue rinsing.
    Dispose of contents/container in accordance with local/regional/national/international regulations.

    Danger

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit