Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ Tetraammineplatinum(II) chloride solution, Pt 9-11% w/w (cont. Pt)

Manufacturer:  Alfa Aesar™ 44045NH

Quantity 1g

 View more versions of this product

Catalog No. AA44045NH

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Specifications Description & Specifications

Chemical Identifiers

13933-32-9
[Pt(NH3)4]Cl2
MFCD00011475
H12Cl2N4Pt
334.11 (Anhydrous)
Tetraammineplatinum (II) chloride solution

Specifications

13933-32-9
H12Cl2N4Pt
334.11 (Anhydrous)
Colorless to pale yellow
1g
Liquid
[Pt(NH3)4]Cl2
MFCD00011475
Pt 9 to 11% w/w (cont. Pt)
Tetraammineplatinum (II) chloride solution

Safety and Handling

SDS & Safety Documents

GHS H Statement
H290-H315-H319
May be corrosive to metals.
Causes skin irritation.
Causes serious eye irritation.

GHS P Statement
P280-P305+P351+P338-P362-P321-P332+P313-P406
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
Continue rinsing.
Take off contaminated clothing and wash before reuse.
Specific treatment (see label).
If skin irritation occurs: Get medical advice/attention.
Store in corrosive resistant container with a resistant inner liner.

Warning

TSCA : TSCA

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit