Alfa Aesar™ Tetraammineplatinum(II) chloride solution, Pt 9-11% w/w (cont. Pt)

Manufacturer: Alfa Aesar™ 44045NB

Quantity 5g

 View more versions of this product

Catalog No. AA44045NB

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 13933-32-9
    Molecular Formula H12Cl2N4Pt
    Linear Formula [Pt(NH3)4]Cl2
    Formula Weight 334.11 (Anhydrous)
    MDL Number MFCD00011475
    Chemical Name or Material Tetraammineplatinum (II) chloride solution

    Specifications

    Formula Weight 334.11 (Anhydrous)
    Health Hazard 3 GHS P Statement
    P280-P305+P351+P338-P362-P321-P332+P313-P406
    Wear protective gloves/protective clothing/eye protection/face protection.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
    Continue rinsing.
    Take off contaminated clothing and wash before reuse.
    Specific treatment (see label).
    If skin irritation occurs: Get medical advice/attention.
    Store in corrosive resistant container with a resistant inner liner.
    MDL Number MFCD00011475
    CAS 13933-32-9
    Health Hazard 1 Warning
    Concentration Pt 9 to 11% w/w (cont. Pt)
    TSCA TSCA
    Physical Form Liquid
    Color Colorless to pale yellow
    Quantity 5g
    Molecular Formula H12Cl2N4Pt
    Linear Formula [Pt(NH3)4]Cl2
    Health Hazard 2 GHS H Statement
    H290-H315-H319
    May be corrosive to metals.
    Causes skin irritation.
    Causes serious eye irritation.
    Chemical Name or Material Tetraammineplatinum (II) chloride solution

    GHS H Statement
    H290-H315-H319
    May be corrosive to metals.
    Causes skin irritation.
    Causes serious eye irritation.

    GHS P Statement
    P280-P305+P351+P338-P362-P321-P332+P313-P406
    Wear protective gloves/protective clothing/eye protection/face protection.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
    Continue rinsing.
    Take off contaminated clothing and wash before reuse.
    Specific treatment (see label).
    If skin irritation occurs: Get medical advice/attention.
    Store in corrosive resistant container with a resistant inner liner.

    Warning

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit