Alfa Aesar™ Tetraammineplatinum(II) hydrogen phosphate solution, Pt 2% w/w (cont. Pt)

Manufacturer: Alfa Aesar™ 44047NB

Quantity 5g

 View more versions of this product

Catalog No. AA44047NB

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 127733-98-6
    Molecular Formula [Pt(NH3)4]HPO4
    Formula Weight 359.18 (Anhydrous)
    MDL Number MFCD03788258
    Synonym 20Q Salt
    Chemical Name or Material Tetraammineplatinum (II) hydrogen phosphate solution

    Specifications

    Formula Weight 359.18 (Anhydrous)
    Health Hazard 3 GHS P Statement
    P284a-P261-P280-P342+P311-P363-P501a
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    Wear protective gloves/protective clothing/eye protection/face protection.
    If experiencing respiratory symptoms: Call a POISON CENTER or doctor/physician.
    Wash contaminated clothing before reuse.
    Dispose of contents/container in accordance with local/regional/national/international regulations.
    Health Hazard 2 GHS H Statement
    H334-H317
    May cause allergy or asthma symptoms or breathing difficulties if inhaled.
    May cause an allergic skin reaction.
    Physical Form Liquid
    MDL Number MFCD03788258
    Synonym 20Q Salt
    Color Colorless
    Concentration Pt 2% w/w (cont. Pt)
    Quantity 5g
    Molecular Formula [Pt(NH3)4]HPO4
    CAS 127733-98-6
    Health Hazard 1 Danger
    Chemical Name or Material Tetraammineplatinum (II) hydrogen phosphate solution

    GHS H Statement
    H334-H317
    May cause allergy or asthma symptoms or breathing difficulties if inhaled.
    May cause an allergic skin reaction.

    GHS P Statement
    P284a-P261-P280-P342+P311-P363-P501a
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    Wear protective gloves/protective clothing/eye protection/face protection.
    If experiencing respiratory symptoms: Call a POISON CENTER or doctor/physician.
    Wash contaminated clothing before reuse.
    Dispose of contents/container in accordance with local/regional/national/international regulations.

    Danger

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit