Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Tetraammineplatinum(II) nitrate solution, Pt 3-4% w/w (cont. Pt), Thermo Scientific Chemicals
Click to view available options
Quantity:
0.5 g
2 g
Description
Tetraammineplatinum(II) nitrate is widely utilized as a precursor to bimetallic Pt/Zn catalysts for the selective hydrogenation of crotonaldehyde.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Color | Colorless |
Physical Form | Liquid |
Quantity | 0.5 g |
Linear Formula | (NH3)4Pt(NO3)2 |
UN Number | UN3218 |
Solubility Information | Miscible with water. |
Formula Weight | 387.21 |
Concentration or Composition (by Analyte or Components) | Pt 3 to 4% w/w (cont. Pt) |
Density | 1.05g/mL |
Chemical Name or Material | Tetraammineplatinum(II) nitrate solution |
Safety and Handling
Hazard Category | Danger |
Hazard Statement | GHS H Statement H240-H272 Heating may cause an explosion. May intensify fire oxidizer. |
Precautionary Statement | GHS P Statement P221-P210-P370+P378a-P410-P411a+P235-P501a Take any precaution to avoid mixing with combustibles. Keep away from heat/sparks/open flames/hot surfaces. No smoking. In case of fire: Use for extinction: CO2, powder or water spray. Protect from sunlight. Store at temperatures not exceeding 392°F. Keep cool. Dispose of contents/container in accordance with local/regional/national/international regulations. |
DOTInformation | Transport Hazard Class: 5.1; Packing Group: III; Proper Shipping Name: NITRATES, INORGANIC, AQUEOUS SOLUTION, N.O.S . |
EINECSNumber | 243-929-9 |
TSCA | No |
Recommended Storage | Ambient temperatures |
RUO – Research Use Only
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction