Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tetrabromophenolphthalein Ethyl Ester, Potassium Salt, Indicator Grade, ACROS Organics™

Click to view available options
Quantity:
1g
5g
25 g
Packaging:
Glass bottle
62637-91-6
C22H13Br4KO4
700.06
MFCD00011662
WCIQBKUTYDIBJC-UHFFFAOYSA-M
23689366
potassium 2,6-dibromo-4-[(3,5-dibromo-4-oxocyclohexa-2,5-dien-1-ylidene)[2-(ethoxycarbonyl)phenyl]methyl]benzen-1-olate
[K+].CCOC(=O)C1=CC=CC=C1C(C1=CC(Br)=C([O-])C(Br)=C1)=C1C=C(Br)C(=O)C(Br)=C1
62637-91-6
C22H13Br4KO4
700.06
MFCD00011662
WCIQBKUTYDIBJC-UHFFFAOYSA-M
23689366
potassium 2,6-dibromo-4-[(3,5-dibromo-4-oxocyclohexa-2,5-dien-1-ylidene)[2-(ethoxycarbonyl)phenyl]methyl]benzen-1-olate
[K+].CCOC(=O)C1=CC=CC=C1C(C1=CC(Br)=C([O-])C(Br)=C1)=C1C=C(Br)C(=O)C(Br)=C1
Indicator
Merck Index 15, 9331
Infrared Spectrum Authentic
Formula Weight 700.05
Physical Form Crystalline Powder or Granules
Grade Extra Pure
Packaging Glass bottle
Specific Extinction 660 min. (600 nm, ethanol)
Color Black-Blue to Blue
Quantity 25 g
Chemical Name or Material Tetrabromophenolphthalein ethyl ester, potassium salt

EINECSNumber : 263-661-6

TSCA : TSCA