Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tetrabromothiophene, 99+%, ACROS Organics™

Click to view available options
Quantity:
25g
3958-03-0
C4Br4S
399.72
MFCD00005419
AVPWUAFYDNQGNZ-UHFFFAOYSA-N
tetrabromothiophene,perbromothiophene,thiophene, tetrabromo,tetrabromthiophen,2,3,4,5-tetrabromo-thiophene,pubchem7661,acmc-1ae1j,2,4,5-tetrabromothiophene,ksc491i3p,2,3,4,5-tetrabromothiophen
77565
2,3,4,5-tetrabromothiophene
C1(=C(SC(=C1Br)Br)Br)Br
3958-03-0
C4Br4S
399.72
MFCD00005419
AVPWUAFYDNQGNZ-UHFFFAOYSA-N
tetrabromothiophene,perbromothiophene,thiophene, tetrabromo,tetrabromthiophen,2,3,4,5-tetrabromo-thiophene,pubchem7661,acmc-1ae1j,2,4,5-tetrabromothiophene,ksc491i3p,2,3,4,5-tetrabromothiophen
77565
2,3,4,5-tetrabromothiophene
C1(=C(SC(=C1Br)Br)Br)Br
Infrared Spectrum Authentic
Color Brown to Cream
Quantity 25g
Melting Point 112.0°C to 116.0°C
Packaging Glass bottle
Boiling Point 326.0°C
Assay Percent Range 99+%
Beilstein 17, 34
Formula Weight 399.71
Physical Form Crystalline Powder
Percent Purity ≥99%
Chemical Name or Material Tetrabromothiophene, 99+%
Show More Show Less

GHS H Statement:
Causes serious eye irritation.
May cause respiratory irritation.
Causes skin irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

EINECSNumber : 223-554-7