Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tetrabutylammonium borohydride, 98%, ACROS Organics™

Click to view available options
Quantity:
10g
50g
Packaging:
Glass bottle
33725-74-5
C16H40BN
257.3
MFCD00012035
SGIQRFZMJGZMNT-UHFFFAOYSA-N
tetrabutylammonium borohydride,tetra-n-butylammonium borohydride,n,n,n-tributylbutan-1-aminium tetrahydroborate,boron 1-; tetrabutylazanium,boron 1-; tetrabutylammonium
9881569
boron(1-);tetrabutylazanium
[B-].CCCC[N+](CCCC)(CCCC)CCCC
33725-74-5
C16H40BN
257.3
MFCD00012035
SGIQRFZMJGZMNT-UHFFFAOYSA-N
tetrabutylammonium borohydride,tetra-n-butylammonium borohydride,n,n,n-tributylbutan-1-aminium tetrahydroborate,boron 1-; tetrabutylazanium,boron 1-; tetrabutylammonium
9881569
boron(1-);tetrabutylazanium
[B-].CCCC[N+](CCCC)(CCCC)CCCC
Reduction reagent
Packaging Glass bottle
Melting Point 126.0°C to 131.0°C
Quantity 10g
Infrared Spectrum Authentic
Color White
Assay Percent Range 98%
Linear Formula [CH3(CH2)3]4NBH4
Fieser 07,352; 10,378; 11,499
Solubility Information Solubility in water: reacts. Other solubilities: soluble in aprotic solvent
Formula Weight 257.3
Physical Form Crystalline Powder and Chunks
Percent Purity 98%
Chemical Name or Material Tetrabutylammonium borohydride, 98%
Show More Show Less

GHS H Statement
Causes severe skin burns and eye damage.
In contact with water releases flammable gas.

GHS P Statement
Do not breathe dust/fume/gas/mist/vapors/spray.
Immediately call a POISON CENTER or doctor/physician.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water

GHS Signal Word: Danger

EINECSNumber : 251-658-2

TSCA : TSCA