Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tetrabutylammonium fluoride, 1M solution in THF, containing ca. 5% water, ACROS Organics™

Click to view available options
Quantity:
5mL
100mL
500mL
2.5L
Packaging:
Glass bottle
429-41-4,109-99-9,7732-18-5
C16H36FN
261.469
MFCD00011747
FPGGTKZVZWFYPV-UHFFFAOYSA-M
tetrabutylammonium fluoride,tbaf,tetrabutylazanium fluoride,tetrabutyl ammonium fluoride,tetra-n-butylammonium fluoride,tetrabutylamine, fluoride,n,n,n-tributylbutan-1-aminium fluoride,1-butanaminium, n,n,n-tributyl-, fluoride,n,n,n-tributyl-1-butanaminium fluoride,1-butanaminium, n,n,n-tributyl-, fluoride 1:1
2724141
CHEBI:51990
tetrabutylazanium;fluoride
CCCC[N+](CCCC)(CCCC)CCCC.[F-]
429-41-4,109-99-9,7732-18-5
C16H36FN
261.469
MFCD00011747
FPGGTKZVZWFYPV-UHFFFAOYSA-M
tetrabutylammonium fluoride,tbaf,tetrabutylazanium fluoride,tetrabutyl ammonium fluoride,tetra-n-butylammonium fluoride,tetrabutylamine, fluoride,n,n,n-tributylbutan-1-aminium fluoride,1-butanaminium, n,n,n-tributyl-, fluoride,n,n,n-tributyl-1-butanaminium fluoride,1-butanaminium, n,n,n-tributyl-, fluoride 1:1
2724141
CHEBI:51990
tetrabutylazanium;fluoride
CCCC[N+](CCCC)(CCCC)CCCC.[F-]
C-X bond formation reagent
Linear Formula [CH3(CH2)3]4NF
Fieser 04,477; 05,645; 07,353; 08,467; 09,444; 10,378; 11,499; 12,458; 14,293; 15,168; 17,363
Density 0.8870g/mL
Formula Weight 261.46
Packaging Glass bottle
Specific Gravity 0.887
Quantity 2.5L
Flash Point −17°C
Chemical Name or Material Tetrabutylammonium fluoride, 1M Solution in THF, Containing ca 5% Water

GHS H Statement
Causes severe skin burns and eye damage. Suspected of causing cancer. May cause respiratory irritation. Highly flammable liquid and vapor. May form explosive peroxides. May cause drowsiness or dizziness.

GHS P Statement
Keep away from heat/sparks/open flames/hot surfaces. - No smoking. IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/shower. Wear protective gloves/protective clothing/eye protection/face protection. IF SWALLOWED: rinse mouth. Do NOT induce vomiting. IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing. Immediately call a POISON CENTER or doctor/physician.

Danger

EINECSNumber : 207-057-2

TSCA : TSCA