Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tetrabutylammonium Hydroxide, 1.0 M Aqueous Solution, HPLC Grade, Spectrum™

Click to view available options
Quantity:
25 mL
500 mL
Packaging:
Amber Glass Bottle
Poly Bottle
2052-49-5,7732-18-5
C16H37NO
259.48
VDZOOKBUILJEDG-UHFFFAOYSA-M
tetrabutylazanium hydroxide
[OH-].CCCC[N+](CCCC)(CCCC)CCCC
2052-49-5,7732-18-5
C16H37NO
259.48
VDZOOKBUILJEDG-UHFFFAOYSA-M
tetrabutylazanium hydroxide
[OH-].CCCC[N+](CCCC)(CCCC)CCCC

T2335, 2052-49-5, C16H37NO

Spectrum™ Tetrabutylammonium Hydroxide, 1.0 M Aqueous Solution, HPLC Grade is a solution for ion pair chromatography. All HPLC components offered by Spectrum are rigorously purified and exacting specification testing to insure accurate, repeatable results.

CAS Max % 0.25,0.75
Percent Purity 1.00 +/-0.02M
Grade HPLC
Packaging Poly Bottle
Quantity 500 mL