Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tetrabutylammonium hydroxide, 40 wt.% (1.5M) solution in water, ACROS Organics™

Click to view available options
Quantity:
50g
250g
1kg
2.5kg
Packaging:
Plastic bottle
2052-49-5,7732-18-5
C16H37NO
259.46
MFCD00009425
VDZOOKBUILJEDG-UHFFFAOYSA-M
tetrabutylammonium hydroxide,tetra-n-butylammonium hydroxide,tetrabutylazanium hydroxide,tetrabutylammoniumhydroxide,1-butanaminium, n,n,n-tributyl-, hydroxide,ammonium, tetrabutyl-, hydroxide,n,n,n-tributyl-1-butanaminium hydroxide,tetra n-butyl ammonium hydroxide,tetra-n-butyl ammonium hydroxide,1-butanaminium, n,n,n-tributyl-, hydroxide 1:1
2723671
tetrabutylazanium;hydroxide
CCCC[N+](CCCC)(CCCC)CCCC.[OH-]
2052-49-5,7732-18-5
C16H37NO
259.46
MFCD00009425
VDZOOKBUILJEDG-UHFFFAOYSA-M
tetrabutylammonium hydroxide,tetra-n-butylammonium hydroxide,tetrabutylazanium hydroxide,tetrabutylammoniumhydroxide,1-butanaminium, n,n,n-tributyl-, hydroxide,ammonium, tetrabutyl-, hydroxide,n,n,n-tributyl-1-butanaminium hydroxide,tetra n-butyl ammonium hydroxide,tetra-n-butyl ammonium hydroxide,1-butanaminium, n,n,n-tributyl-, hydroxide 1:1
2723671
tetrabutylazanium;hydroxide
CCCC[N+](CCCC)(CCCC)CCCC.[OH-]
Phase transfer catalyst
Color White to Yellow
Specific Gravity 0.995
Density 0.9950g/mL
Boiling Point >100.0°C
Packaging Plastic bottle
Quantity 250g
Linear Formula [CH3(CH2)3]4NOH
Beilstein 04, II, 634
Fieser 05,645; 11,500
Solubility Information Solubility in water: soluble.
Formula Weight 259.46
Physical Form Powder or Crystals
Percent Purity 38 to 42% (Total base)
Chemical Name or Material Tetrabutylammonium hydroxide, 40 wt.% Solution in Water
Show More Show Less

GHS H Statement
Causes severe skin burns and eye damage.
Harmful if swallowed.

GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: Rinse mouth.
Do NOT induce vomiting.
IF ON SKIN (or hair): Take off immediately all contaminated clothing.
Rinse skin with wat

GHS Signal Word: Danger

EINECSNumber : 218-147-6