Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tetrabutylammonium hydroxide, 40 wt.% solution in methanol, ACROS Organics™

Click to view available options
Quantity:
100mL
500mL
2.5L
Packaging:
Glass bottle
2052-49-5,67-56-1
C16H37NO
259.46
MFCD00009425
VDZOOKBUILJEDG-UHFFFAOYSA-M
tetrabutylammonium hydroxide,tetra-n-butylammonium hydroxide,tetrabutylazanium hydroxide,tetrabutylammoniumhydroxide,1-butanaminium, n,n,n-tributyl-, hydroxide,ammonium, tetrabutyl-, hydroxide,n,n,n-tributyl-1-butanaminium hydroxide,tetra n-butyl ammonium hydroxide,tetra-n-butyl ammonium hydroxide,1-butanaminium, n,n,n-tributyl-, hydroxide 1:1
2723671
tetrabutylazanium;hydroxide
CCCC[N+](CCCC)(CCCC)CCCC.[OH-]
2052-49-5,67-56-1
C16H37NO
259.46
MFCD00009425
VDZOOKBUILJEDG-UHFFFAOYSA-M
tetrabutylammonium hydroxide,tetra-n-butylammonium hydroxide,tetrabutylazanium hydroxide,tetrabutylammoniumhydroxide,1-butanaminium, n,n,n-tributyl-, hydroxide,ammonium, tetrabutyl-, hydroxide,n,n,n-tributyl-1-butanaminium hydroxide,tetra n-butyl ammonium hydroxide,tetra-n-butyl ammonium hydroxide,1-butanaminium, n,n,n-tributyl-, hydroxide 1:1
2723671
tetrabutylazanium;hydroxide
CCCC[N+](CCCC)(CCCC)CCCC.[OH-]
Phase transfer catalyst
Specific Gravity 0.91
Density 0.9100g/mL
Boiling Point 65.0°C
Flash Point 12°C
Quantity 500mL
Color Colorless to Yellow
Melting Point -98.0°C
Packaging Glass bottle
Linear Formula [CH3(CH2)3]4NOH
Beilstein 04, II, 634
Fieser 05,645; 11,500
Solubility Information Solubility in water: soluble
Formula Weight 259.46
Physical Form Solution
Percent Purity ≥40%
Chemical Name or Material Tetrabutylammonium hydroxide, 40 wt.% solution in methanol
Show More Show Less

GHS H Statement
Toxic in contact with skin.
Toxic if swallowed.
Causes severe skin burns and eye damage.
Toxic if inhaled.
Causes damage to organs.
Highly flammable liquid and vapor.

GHS P Statement
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove conta

GHS Signal Word: Danger

EINECSNumber : 218-147-6