Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tetrabutylammonium Hydroxide Solution, 1.0M, Honeywell Fluka™

Click to view available options
Quantity:
100mL
500mL
2052-49-5
C16H37NO
259.478
MFCD00009425
VDZOOKBUILJEDG-UHFFFAOYSA-M
tetrabutylammonium hydroxide,tetra-n-butylammonium hydroxide,tetrabutylazanium hydroxide,tetrabutylammoniumhydroxide,1-butanaminium, n,n,n-tributyl-, hydroxide,ammonium, tetrabutyl-, hydroxide,n,n,n-tributyl-1-butanaminium hydroxide,tetra n-butyl ammonium hydroxide,tetra-n-butyl ammonium hydroxide,1-butanaminium, n,n,n-tributyl-, hydroxide 1:1
2723671
tetrabutylazanium;hydroxide
CCCC[N+](CCCC)(CCCC)CCCC.[OH-]
2052-49-5
C16H37NO
259.478
MFCD00009425
VDZOOKBUILJEDG-UHFFFAOYSA-M
tetrabutylammonium hydroxide,tetra-n-butylammonium hydroxide,tetrabutylazanium hydroxide,tetrabutylammoniumhydroxide,1-butanaminium, n,n,n-tributyl-, hydroxide,ammonium, tetrabutyl-, hydroxide,n,n,n-tributyl-1-butanaminium hydroxide,tetra n-butyl ammonium hydroxide,tetra-n-butyl ammonium hydroxide,1-butanaminium, n,n,n-tributyl-, hydroxide 1:1
2723671
tetrabutylazanium;hydroxide
CCCC[N+](CCCC)(CCCC)CCCC.[OH-]
Quantity 100mL
Linear Formula (CH3CH2CH2CH2)4N(OH)
UN Number UN3267
Beilstein 3571228
Formula Weight 259.47g/mol
Chemical Name or Material Tetrabutylammonium hydroxide solution

P280-P305 + P351 + P338-P310