Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Tetrabutylammonium Phosphate (White to Off-White Powder/HPLC Grade), Fisher Chemical™
Click to view available options
Quantity:
5 g
25 g
100 g
Description
- High purity
- Application tested
- Dissolves easily
Specifications
Absorbance | 0.02 AU max. at 214nm, 0.1 AU max. at 210nm, 0.2 AU max. at 200nm |
Melting Point | 148°C |
Color | White |
pH | 4 to 5 |
Assay Percent Range | ≥98.5 % |
Packaging | Glass Bottle |
Linear Formula | (CH3CH2CH2CH2)4NH2PO4 |
Quantity | 5 g |
Formula Weight | 339.45 |
Percent Purity | ≥98.5% |
Show More |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction