Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tetrabutylammonium Tetrafluoroborate 98.0+%, TCI America™

Click to view available options
Quantity:
25g
100g
500g
429-42-5
C16H36BF4N
329.275
MFCD00011634
NNZZSJSQYOFZAM-UHFFFAOYSA-N
tetrabutylammonium tetrafluoroborate,tetra-n-butylammonium tetrafluoroborate,tetrabutylammonium fluoroborate,1-butanaminium, n,n,n-tributyl-, tetrafluoroborate 1-,tetrabutyl ammonium tetrafluoroborate,tetrabutylammonium ion tetrafluoroborate,1-butanaminium, n,n,n-tributyl-, tetrafluoroborate 1-1:1,tbabf4,acmc-209jry,tbabf4;
67932
tetrabutylazanium;tetrafluoroborate
[B-](F)(F)(F)F.CCCC[N+](CCCC)(CCCC)CCCC
429-42-5
C16H36BF4N
329.275
MFCD00011634
NNZZSJSQYOFZAM-UHFFFAOYSA-N
tetrabutylammonium tetrafluoroborate,tetra-n-butylammonium tetrafluoroborate,tetrabutylammonium fluoroborate,1-butanaminium, n,n,n-tributyl-, tetrafluoroborate 1-,tetrabutyl ammonium tetrafluoroborate,tetrabutylammonium ion tetrafluoroborate,1-butanaminium, n,n,n-tributyl-, tetrafluoroborate 1-1:1,tbabf4,acmc-209jry,tbabf4;
67932
tetrabutylazanium;tetrafluoroborate
[B-](F)(F)(F)F.CCCC[N+](CCCC)(CCCC)CCCC
Quantity 100g
Color White
Melting Point 161°C
UN Number 1759
Formula Weight 329.27
Physical Form Crystal-Powder at 20°C
Percent Purity ≥98.0% (N)
Chemical Name or Material Tetrabutylammonium Tetrafluoroborate

EINECSNumber : (2)-0186

TSCA : Yes