Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tetrabutylphosphonium Chloride (80% in Water), TCI America™

Click to view available options
Quantity:
25g
2304-30-5
C16H36ClP
294.888
MFCD00011854
IBWGNZVCJVLSHB-UHFFFAOYSA-M
75311
tetrabutylphosphanium;chloride
CCCC[P+](CCCC)(CCCC)CCCC.[Cl-]
2304-30-5
C16H36ClP
294.888
MFCD00011854
IBWGNZVCJVLSHB-UHFFFAOYSA-M
75311
tetrabutylphosphanium;chloride
CCCC[P+](CCCC)(CCCC)CCCC.[Cl-]
Quantity 25g
Color Yellow
UN Number 3278
Formula Weight 294.89
Physical Form Clear Liquid at 20°C
Chemical Name or Material Tetrabutylphosphonium Chloride (80% in Water)

RTECSNumber : TA2419000

TSCA : Yes