Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tetracaine Hydrochloride, USP, 98-102%, Spectrum™

Click to view available options
Quantity:
25 g
100 g
1 kg
5 kg
25 kg
Packaging:
Amber Glass Bottle
Fiber Drum
Poly Pail
136-47-0
C15H25ClN2O2
300.83
PPWHTZKZQNXVAE-UHFFFAOYSA-N
hydrogen 2-(dimethylamino)ethyl 4-(butylamino)benzoate chloride
[H+].[Cl-].CCCCNC1=CC=C(C=C1)C(=O)OCCN(C)C
136-47-0
C15H25ClN2O2
300.83
PPWHTZKZQNXVAE-UHFFFAOYSA-N
hydrogen 2-(dimethylamino)ethyl 4-(butylamino)benzoate chloride
[H+].[Cl-].CCCCNC1=CC=C(C=C1)C(=O)OCCN(C)C

TE128, 136-47-0, C15H24N2O2•HCl

Spectrum™ Tetracaine Hydrochloride, USP is a local anesthetic used to prevent pain and induce anesthesia. All Spectrum Chemical USP products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.

Packaging Amber Glass Bottle
Ignition Residue 0.001
Water 0.02
Quantity 100 g
CAS Max % 1
Percent Purity 98 to 102%
Grade USP