Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tetracaine, USP, 98-102%, Spectrum™

Click to view available options
Quantity:
5 g
25 g
100 g
500 g
1 kg
5 kg
20 kg
Packaging:
Amber Glass Bottle
Poly Pail
94-24-6
C15H24N2O2
264.37
GKCBAIGFKIBETG-UHFFFAOYSA-N
2-(dimethylamino)ethyl 4-(butylamino)benzoate
CCCCNC1=CC=C(C=C1)C(=O)OCCN(C)C
94-24-6
C15H24N2O2
264.37
GKCBAIGFKIBETG-UHFFFAOYSA-N
2-(dimethylamino)ethyl 4-(butylamino)benzoate
CCCCNC1=CC=C(C=C1)C(=O)OCCN(C)C

TE220, 94-24-6, C15H24N2O2

Spectrum™ Tetracaine, USP is a local anesthetic used to prevent pain and induce anesthesia. All Spectrum Chemical USP products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.

Packaging Amber Glass Bottle
Loss on Drying 0.005
Ignition Residue 0.001
Quantity 1 kg
CAS Max % 1
Percent Purity 98 to 102%
Grade USP