Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tetracarbonyldi-µ-chlorodirhodium(I), 97%, ACROS Organics™

Click to view available options
Quantity:
250mg
1g
Packaging:
Glass bottle
14523-22-9
C4Cl2O4Rh2
388.76
MFCD00135610
FGKDXBICTVUSPK-UHFFFAOYSA-L
tetracarbonyldi-mu-chlorodirhodium i,rhodium carbonyl chloride i,rhodium i carbonyl chloride,rhodium i dicarbonyl chloride dimer,di-mu-chloro-tetracarbonyldirhodium i,chlorodicarbonylrhodium i dimer,4co.cl2rh2,bis chlorodicarbonyl rhodium,tetracarbonyldi-,i-chlorodirhodium i,tetracarbonyldi-micron-chlorodirhodium i
14523-22-9
C4Cl2O4Rh2
388.76
MFCD00135610
FGKDXBICTVUSPK-UHFFFAOYSA-L
tetracarbonyldi-mu-chlorodirhodium i,rhodium carbonyl chloride i,rhodium i carbonyl chloride,rhodium i dicarbonyl chloride dimer,di-mu-chloro-tetracarbonyldirhodium i,chlorodicarbonylrhodium i dimer,4co.cl2rh2,bis chlorodicarbonyl rhodium,tetracarbonyldi-,i-chlorodirhodium i,tetracarbonyldi-micron-chlorodirhodium i
Packaging Glass bottle
Quantity 1g
Infrared Spectrum Authentic
Melting Point 121.0°C
Color Brown to Red
Assay Percent Range 96% min. (ex Rh)
Linear Formula [Rh(CO)2Cl]2
UN Number 2811
Merck Index 15, 8312
Solubility Information Solubility in water: insoluble. Other solubilities: soluble in most common organic solvents (except, aliphatic hydrocarbons)
Formula Weight 388.76
Physical Form Crystals, Crystalline Powder or Needles
Percent Purity 97%
Chemical Name or Material Tetracarbonyldi-µ-chlorodirhodium(I)
Show More Show Less

GHS H Statement
Harmful if swallowed.

GHS P Statement
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
Wash face,hands and any exposed skin thoroughly after handling.
Wear protective gloves/protective clothing/eye protection/face protection.

GHS Signal Word: Warning

EINECSNumber : 238-540-6