Tetracarbonyldi-mu-chlorodirhodium(I), Rh 50.1-52.9%, Alfa Aesar™

Manufacturer: Alfa Aesar 1054701

Quantity 0.25g

 View more versions of this product

Catalog No. AA1054701

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 14523-22-9
    Physical Form Crystalline
    Molecular Formula [RhCl(CO)2]2
    Formula Weight 388.76
    MDL Number MFCD00135610
    Synonym Rhodium carbonyl chloride
    Chemical Name or Material Tetracarbonyldi-mu-chlorodirhodium(I), Rh 50.1-52.9%

    Specifications

    Formula Weight 388.76
    Quantity 0.25g
    EINECS Number 238-540-6
    Physical Form Crystalline
    Molecular Formula [RhCl(CO)2]2
    MDL Number MFCD00135610
    CAS 14523-22-9
    Synonym Rhodium carbonyl chloride
    Chemical Name or Material Tetracarbonyldi-mu-chlorodirhodium(I), Rh 50.1-52.9%

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit