Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ Tetrachlorobis(tetrahydrofuran)niobium(IV)

Manufacturer:  Alfa Aesar™ 3946009

Quantity 10g

 View more versions of this product

Catalog No. AA3946009

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Specifications Description & Specifications

Chemical Identifiers

61247-57-2
MFCD00013405
Niobium(IV) chloride - tetrahydrofuran complex
378.94
UN2925
Tetrachlorobis(tetrahydrofuran)niobium(IV)

Specifications

61247-57-2
378.94
UN2925
Yellow-orange to brown
49° (120°F)
Moisture Sensitive
Powder
MFCD00013405
Niobium(IV) chloride - tetrahydrofuran complex
93°C (dec.)
10g
Tetrachlorobis(tetrahydrofuran)niobium(IV)

Safety and Handling

SDS & Safety Documents

GHS H Statement
H228-H314-H318
Flammable solid.
Causes severe skin burns and eye damage.
Causes serious eye damage.

GHS P Statement
P210-P260-P303+P361+P353-P305+P351+P338-P405-P501a
Keep away from heat/sparks/open flames/hot surfaces.
No smoking.
Do not breathe dust/fume/gas/mist/vapors/spray.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing.
Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Danger

DOTInformation : Transport Hazard Class: 4.1; Packing Group: II; Proper Shipping Name: FLAMMABLE SOLIDS, CORROSIVE, ORGANIC, N.O.S.

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit