Alfa Aesar™ Tetrachlorobis(tetrahydrofuran)titanium(IV), 98%

Manufacturer: Alfa Aesar™ 3959404

Quantity 2g

 View more versions of this product

Catalog No. AA3959404

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 31011-57-1
    Formula Weight 333.91
    MDL Number MFCD00077884
    UN Number UN2925
    Synonym Titanium(IV) chloride - tetrahydrofuran complex
    Chemical Name or Material Tetrachlorobis(tetrahydrofuran)titanium(IV)

    Specifications

    Physical Form Powder/Crystalline
    Formula Weight 333.91
    Health Hazard 3 GHS P Statement
    P210-P260-P303+P361+P353-P305+P351+P338-P405-P501a
    Keep away from heat/sparks/open flames/hot surfaces.
    No smoking.
    Do not breathe dust/fume/gas/mist/vapors/spray.
    IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing.
    Rinse skin with water/shower.
    IF IN EYES: Rinse cautiously with water for several minutes.
    Remove contact lenses, if present and easy to do.
    Continue rinsing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.
    CAS 31011-57-1
    Health Hazard 2 GHS H Statement
    H228-H314-H318
    Flammable solid.
    Causes severe skin burns and eye damage.
    Causes serious eye damage.
    Synonym Titanium(IV) chloride - tetrahydrofuran complex
    Health Hazard 1 Danger
    Quantity 2g
    UN Number UN2925
    Color Yellow
    Melting Point 126° to 128°C
    Flash Point −17° (1°F)
    Sensitivity Moisture Sensitive
    MDL Number MFCD00077884
    DOT Information Transport Hazard Class: 4.1; Packing Group: II; Proper Shipping Name: FLAMMABLE SOLIDS, CORROSIVE, ORGANIC, N.O.S.
    Chemical Name or Material Tetrachlorobis(tetrahydrofuran)titanium(IV)
    Assay 98%

    GHS H Statement
    H228-H314-H318
    Flammable solid.
    Causes severe skin burns and eye damage.
    Causes serious eye damage.

    GHS P Statement
    P210-P260-P303+P361+P353-P305+P351+P338-P405-P501a
    Keep away from heat/sparks/open flames/hot surfaces.
    No smoking.
    Do not breathe dust/fume/gas/mist/vapors/spray.
    IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing.
    Rinse skin with water/shower.
    IF IN EYES: Rinse cautiously with water for several minutes.
    Remove contact lenses, if present and easy to do.
    Continue rinsing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.

    Danger

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit