Tetrachloroethylene, 99%

Manufacturer: Alfa Aesar B200890F

Quantity 2500mL

 View more versions of this product

Catalog No. AAB200890F

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 127-18-4
    Molecular Formula Cl2=CCl2
    Linear Formula 2C=CCl2
    Formula Weight 165.83
    MDL Number MFCD00000834
    UN Number UN1897
    Beilstein 1361721
    Merck Index 14,9190
    Chemical Name or Material Tetrachloroethylene

    Specifications

    Formula Weight 165.83
    Quantity 2500mL
    TSCA TSCA
    Linear Formula 2C=CCl2
    MDL Number MFCD00000834
    UN Number UN1897
    Beilstein 1361721
    Merck Index 14,9190
    Health Hazard 1 Warning
    Health Hazard 2 GHS H Statement
    H351
    Suspected of causing cancer.
    Health Hazard 3 GHS P Statement
    P281-P201-P202-P308+P313-P405-P501a
    Use personal protective equipment as required.
    Obtain special instructions before use.
    Do not handle until all safety precautions have been read and understood.
    IF exposed or c
    DOT Information Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: TETRACHLOROETHYLENE
    RTECS Number KX3850000
    Physical Form Liquid
    Density 1.623
    Assay Percent Range 0.99
    Molecular Formula Cl2=CCl2
    CAS 127-18-4
    Chemical Name or Material Tetrachloroethylene
    Assay 99%

    Description

    Applications
    Tetrachloroethylene is used as a solvent. It is involved in dry cleaning as well as in degreasing of metal parts. It finds application as a neutrino detector. It is utilized as a paint stripper and as a spot remover in consumer products. It acts as a precursor for the manufacture of other chemicals. It plays an important role in water repellants, paint removers, printing inks, glues, sealants, polishes and lubricants. As a film-forming electrolyte additive, it is used in the manufacture of lithium ion batteries and is actively involved as an extraction solvent in the determination of oil and grease in water by FT-IR.

    Solubility
    Miscible with alcohol, ether, chloroform, benzene and hexane. Slightly miscible with water.

    Notes
    Incompatible with strong oxidizing agents, strong bases, alkali metals and aluminum.

    GHS H Statement
    H351
    Suspected of causing cancer.

    GHS P Statement
    P281-P201-P202-P308+P313-P405-P501a
    Use personal protective equipment as required.
    Obtain special instructions before use.
    Do not handle until all safety precautions have been read and understood.
    IF exposed or c

    Warning

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit