Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tetrachloroethylene, BAKER ANALYZED™ Reagent, J.T.Baker™

Click to view available options
Quantity:
4L
500mL
Packaging:
Narrow Mouth Amber Glass Bottle

Chemical Identifiers

127-18-4
C2Cl4
165.822
CYTYCFOTNPOANT-UHFFFAOYSA-N
31373
CHEBI:17300

High quality chemicals for laboratory and specialized industrial use

CAS 127-18-4
Molecular Formula C2Cl4
InChI Key CYTYCFOTNPOANT-UHFFFAOYSA-N
SMILES C(=C(Cl)Cl)(Cl)Cl
Packaging Narrow Mouth Amber Glass Bottle
IUPAC Name 1,1,2,2-tetrachloroethene
Molecular Weight (g/mol) 165.822
PubChem CID 31373
ChEBI CHEBI:17300
Formula Weight 165.83
Specific Gravity 1L = 1.62kg
Quantity 500mL
Chemical Name or Material Tetrachloroethylene
Grade BAKER ANALYZED™ Reagent
Show More Show Less