Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tetrachloroethylene, BAKER ANALYZED™ Reagent, J.T.Baker™

Click to view available options
Quantity:
500mL
4L
Packaging:
Narrow Mouth Amber Glass Bottle

Chemical Identifiers

127-18-4
C2Cl4
165.822
CYTYCFOTNPOANT-UHFFFAOYSA-N
31373
CHEBI:17300
1,1,2,2-tetrachloroethene
C(=C(Cl)Cl)(Cl)Cl

High quality chemicals for laboratory and specialized industrial use

Packaging Narrow Mouth Amber Glass Bottle
Formula Weight 165.83
Specific Gravity 1L = 1.62kg
Quantity 4L
Chemical Name or Material Tetrachloroethylene
Grade BAKER ANALYZED™ Reagent